
(2-Methoxy-6-(trifluoromethyl)phenyl)acetic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1017779-10-0
:
Formula:C₁₀H₉F₃O₃
MDL Number:MFCD09832328
:234.17 g/mol
Melting Pt:148…150 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together