2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1323966-11-5
:
Formula:C₁₀H₇F₄N
MDL Number:MFCD19687199
:217.16 g/mol
Storage Temperature:Ambient
Frequently Bought Together