Order Entry
Czech Republic
ContactUsLinkComponent
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoyl chloride 97%
Corrosive
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoyl chloride 97%
Catalog # APOSPC302805-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoyl chloride 97%
Catalog # APOSPC302805-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.58 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
 :GHS05
CAS Number:1373920-78-5
Formula:C₉H₅ClF₄O₂
MDL Number:MFCD22201039