Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2',4'-Difluoro-3'-methoxyacetophenone
2',4'-Difluoro-3'-methoxyacetophenone 97%
2',4'-Difluoro-3'-methoxyacetophenone 97%
APOSPC302661-1G
:
Apollo Scientific
:
2',4'-Difluoro-3'-methoxyacetophenone 97%
APOSPC302661-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
186.15 g/mol
:
Ambient
:
373603-19-1
:
C₉H₈F₂O₂
:
MFCD02258906