
6-Chloro-2-fluoro-3-methoxybenzyl bromide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886499-73-6
:
Formula:C₈H₇BrClFO
MDL Number:MFCD04115950
:253.50 g/mol
Melting Pt:47…50 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together