
4-Methoxy-2-(trifluoromethoxy)benzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886502-37-0
:
Formula:C₉H₇F₃O₄
MDL Number:MFCD06660285
:236.15 g/mol
Melting Pt:99…101 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together