
2-Chloro-6-fluoro-3-methoxybenzyl bromide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886499-54-3
:
Formula:C₈H₇BrClFO
MDL Number:MFCD04115945
:253.50 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together