Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-2-(trifluoromethoxy)benzamide
5-Chloro-2-(trifluoromethoxy)benzamide 98%
5-Chloro-2-(trifluoromethoxy)benzamide 98%
APOSPC302612-1G
:
Apollo Scientific
:
5-Chloro-2-(trifluoromethoxy)benzamide 98%
APOSPC302612-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
239.58 g/mol
:
Ambient
:
C₈H₅ClF₃NO₂
:
MFCD11519356