
(4-Methoxy-3-(trifluoromethyl)phenyl)acetic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000566-45-9
:
Formula:C₁₀H₉F₃O₃
MDL Number:MFCD09832302
:234.17 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together