4-Methoxy-2-(trifluoromethoxy)benzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886502-33-6
:
Formula:C₉H₆F₃NO₂
MDL Number:MFCD06660284
:217.1447 g/mol
Melting Pt:31…33 °C
Storage Temperature:Ambient
Frequently Bought Together