Order Entry
Czech Republic
ContactUsLinkComponent
4-Ethoxy-2-fluoroaniline 98%
4-Ethoxy-2-fluoroaniline 98%
Catalog # APOSPC302585-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Ethoxy-2-fluoroaniline 98%
Catalog # APOSPC302585-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :155.17 g/mol
Storage Temperature:Ambient
CAS Number:470702-37-5
Formula:C₈H₁₀FNO
MDL Number:MFCD11053973