4-(Difluoromethoxy)-2-fluorobenzamide 97%
Catalog # APOSPC302559-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.14 g/mol
Storage Temperature:Ambient
Formula:C₈H₆F₃NO₂
MDL Number:MFCD11519384