Order Entry
Czech Republic
ContactUsLinkComponent
3-(4-Fluoro-3-(trifluoromethoxy)phenyl)propionic acid 98%
3-(4-Fluoro-3-(trifluoromethoxy)phenyl)propionic acid 98%
Katalogové číslo APOSPC302515-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Fluoro-3-(trifluoromethoxy)phenyl)propionic acid 98%
Katalogové číslo APOSPC302515-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :252.16 g/mol
Storage Temperature:Ambient
CAS Number:1240256-80-7
Formula:C₁₀H₈F₄O₃
MDL Number:MFCD09999449