3-(3-Fluoro-5-(trifluoromethoxy)phenyl)propionic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1261828-37-8
:
Formula:C₁₀H₈F₄O₃
MDL Number:MFCD18783232
:252.16 g/mol
Storage Temperature:Ambient
Frequently Bought Together