Order Entry
Czech Republic
ContactUsLinkComponent
3-(3-(Difluoromethoxy)phenyl)propionic acid 98%
3-(3-(Difluoromethoxy)phenyl)propionic acid 98%
Catalog # APOSPC302504-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(3-(Difluoromethoxy)phenyl)propionic acid 98%
Catalog # APOSPC302504-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :216.18 g/mol
Storage Temperature:Ambient
CAS Number:1092460-66-6
Formula:C₁₀H₁₀F₂O₃
MDL Number:MFCD11519314