Order Entry
Czech Republic
ContactUsLinkComponent
3-(4-Chloro-3-ethoxy-2-fluorophenyl)propionic acid 97%
Irritant
3-(4-Chloro-3-ethoxy-2-fluorophenyl)propionic acid 97%
Catalog # APOSPC302487-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(4-Chloro-3-ethoxy-2-fluorophenyl)propionic acid 97%
Catalog # APOSPC302487-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :246.66 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1323966-45-5
Formula:C₁₁H₁₂ClFO₃
MDL Number:MFCD19687220