0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
3-(2-Fluoro-4-methylphenyl)propionic acid
3-(2-Fluoro-4-methylphenyl)propionic acid 97%
3-(2-Fluoro-4-methylphenyl)propionic acid 97%
APOSPC302476-1G
:
Apollo Scientific
:
3-(2-Fluoro-4-methylphenyl)propionic acid 97%
APOSPC302476-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
182.19 g/mol
:
98…100 °C
:
GHS07
:
Ambient
:
C₁₀H₁₁FO₂
:
MFCD09025394