
(2-Methoxy-4-(trifluoromethyl)phenyl)acetic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1017779-22-4
:
Formula:C₁₀H₉F₃O₃
MDL Number:MFCD09832353
:234.17 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together