2-Fluoro-5-methoxy-3-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1373920-67-2
:
Formula:C₉H₆F₄O₂
MDL Number:MFCD22201033
:222.14 g/mol
Storage Temperature:Ambient
Frequently Bought Together