
2-Chloro-6-(trifluoromethoxy)benzonitrile
Catalog # APOSPC302444-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.56 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1261779-40-1
Formula:C₈H₃ClF₃NO
MDL Number:MFCD18393805