2-(Trifluoromethoxy)-DL-phenylalanine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1218228-20-6
:
Formula:C₁₀H₁₀F₃NO₃
MDL Number:MFCD07371965
:249.19 g/mol
Melting Pt:236 °C
Storage Temperature:Ambient
Frequently Bought Together