
2-(Difluoromethoxy)-5-fluorobenzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1017778-48-1
:
Formula:C₈H₄F₃NO
MDL Number:MFCD09832329
:187.12 g/mol
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
Frequently Bought Together