Order Entry
Czech Republic
ContactUsLinkComponent
3-(Difluoromethoxy)cinnamic acid 97%
3-(Difluoromethoxy)cinnamic acid 97%
Catalog # APOSPC302267-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Difluoromethoxy)cinnamic acid 97%
Catalog # APOSPC302267-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.17 g/mol
Storage Temperature:Ambient
CAS Number:630424-87-2
Formula:C₁₀H₈F₂O₃
MDL Number:MFCD06205319