2-Methoxy-5-(pentafluorothio)aniline 98%
Catalog # APOSPC302255-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:249.20 g/mol
Storage Temperature:Ambient
CAS Number:1211587-84-6
Formula:C₇H₈F₅NOS
MDL Number:MFCD16652485