2,3-Dichloro-6-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1017777-92-2
:
Formula:C₇H₄Cl₂F₃N
MDL Number:MFCD09832277
:230.02 g/mol
Melting Pt:43…45 °C
Storage Temperature:Ambient
Frequently Bought Together