Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Ethoxy-3-(trifluoromethyl)benzamide
4-Ethoxy-3-(trifluoromethyl)benzamide 98%
4-Ethoxy-3-(trifluoromethyl)benzamide 98%
APOSPC302210-1G
:
Apollo Scientific
:
4-Ethoxy-3-(trifluoromethyl)benzamide 98%
APOSPC302210-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
233.19 g/mol
:
Ambient
:
1206593-26-1
:
C₁₀H₁₀F₃NO₂
:
MFCD13196583