
2'-Methyl-4'-(trifluoromethoxy)acetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1373920-81-0
:
Formula:C₁₀H₉F₃O₂
MDL Number:MFCD22201061
:218.17 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together