4-n-Butoxy-2,6-difluorobenzoyl chloride 97%
Catalog # APOSPC302101-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:248.65 g/mol
Storage Temperature:Ambient
CAS Number:1373920-74-1
Formula:C₁₁H₁ClF₂O
MDL Number:MFCD22201027