Order Entry
Czech Republic
ContactUsLinkComponent
4-(Dimethylamino)-3-nitrobenzotrifluoride 97%
Irritant
4-(Dimethylamino)-3-nitrobenzotrifluoride 97%
Catalog # APOSPC3009M-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(Dimethylamino)-3-nitrobenzotrifluoride 97%
Catalog # APOSPC3009M-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Flash Pt:>110 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:40700-38-7
Formula:C₉H₉F₃N₂O₂
MDL Number:MFCD00236670