0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol
APOSPC300994-1G
:
Apollo Scientific
:
1-(2-Fluoro-1,1'-biphenyl-4-yl)ethanol
APOSPC300994-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
216.26 g/mol
:
GHS07
:
Ambient
:
56430-67-2
:
C₁₄H₁₃FO
:
MFCD18089274