Order Entry
Czech Republic
ContactUsLinkComponent
3-(Dimethylamino)-1-(2-fluoro-1,1'-biphenyl-4-yl)prop-2-en-1-one
3-(Dimethylamino)-1-(2-fluoro-1,1'-biphenyl-4-yl)prop-2-en-1-one
Catalog # APOSPC300992-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Dimethylamino)-1-(2-fluoro-1,1'-biphenyl-4-yl)prop-2-en-1-one
Catalog # APOSPC300992-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :269.32 g/mol
Storage Temperature:Ambient
CAS Number:1287227-88-6
Formula:C₁₇H₁₆FNO
MDL Number:MFCD18089276