
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₉H₄ClF₃N₂O₂·xH₂O
MDL Number:MFCD22682820
:264.59 g/mol (anhy)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together