
4-Iodo-3-(trifluoromethyl)-1-H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:866638-72-4
:
Formula:C₄H₂F₃IN₂
MDL Number:MFCD08064883
:261.974 g/mol
Melting Pt:137…141 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together