4-Phenoxy-2-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:106877-21-8
:
Formula:C₁₃H₁₀F₃NO
MDL Number:MFCD08688475
:253.22 g/mol
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
Frequently Bought Together