
4-(4-Trifluoromethyl-2-pyrimidinyloxy)benzenesulphonyl chloride
Catalog # APOSPC300600-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:338.685 g/mol
Melting Pt:108…110 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:1160058-86-5
Formula:C₁₁H₆ClF₃N₂O₃S
MDL Number:MFCD11109532