Order Entry
Czech Republic
ContactUsLinkComponent
 
3-(Dimethylamino)-1-(4-fluorophenyl)-2-propen-1-one
Catalog # APOSPC300544-5G
Supplier:  Apollo Scientific
CAS Number:  
3-(Dimethylamino)-1-(4-fluorophenyl)-2-propen-1-one
Catalog # APOSPC300544-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :193.22 g/mol
Storage Temperature:Ambient
CAS Number:75175-77-8
Formula:C₁₁H₁₂FNO
MDL Number:MFCD00111509