Order Entry
Czech Republic
ContactUsLinkComponent
1-(3-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Catalog # APOSPC300524-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid
Catalog # APOSPC300524-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.21 g/mol
Storage Temperature:Ambient
CAS Number:566154-63-0
Formula:C₁₁H₁₀FNO₃
MDL Number:MFCD01569310