
1,4-Dimethoxy-2-fluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:82830-49-7
:
Formula:C₈H₉FO₂
MDL Number:MFCD00042487
:156.16 g/mol
Boiling Pt:119…121 °C/40mmHg
Melting Pt:23…26 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together