Order Entry
Czech Republic
ContactUsLinkComponent
5-Bromo-2-fluoro-3-methylpyridine
Irritant
5-Bromo-2-fluoro-3-methylpyridine
Catalog # APOSPC2989-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-2-fluoro-3-methylpyridine
Catalog # APOSPC2989-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.01 g/mol
Melting Pt:60…65 °C
Density:1.6 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:29312-98-9
Formula:C₆H₅BrFN
MDL Number:MFCD03095091