
5-Fluoro-2-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:827-20-3
:
Formula:C₇H₅F₄N
MDL Number:MFCD06660353
:179.12 g/mol
Melting Pt:29…32 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together