
1,8-Diiodohexadecafluorooctane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:335-70-6
:
Formula:C₈F₁₆I₂
MDL Number:MFCD00236093
:653.87 g/mol
Boiling Pt:112 °C/25mmHg
Melting Pt:75…76 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together