Methyl-4-hydroxy-3-(trifluoromethyl)benzoate 98+%
Catalog # APOSPC2901-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:220.1500 g/mol
Storage Temperature:Ambient
CAS Number:115933-50-1
Formula:C₉H₇F₃O₃
MDL Number:MFCD06204091