Order Entry
Czech Republic
ContactUsLinkComponent
4-(2,4-Difluorophenyl)-3-thiosemicarbazide 97%
4-(2,4-Difluorophenyl)-3-thiosemicarbazide 97%
Catalog # APOSPC2875W-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(2,4-Difluorophenyl)-3-thiosemicarbazide 97%
Catalog # APOSPC2875W-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:172…174 °C
Storage Temperature:Ambient
CAS Number:206559-58-2
Formula:C₇H₇F₂N₃S
MDL Number:MFCD00060563