Order Entry
Czech Republic
ContactUsLinkComponent
2,4-Difluorophenyl isothiocyanate 97%
Irritant
2,4-Difluorophenyl isothiocyanate 97%
Catalog # APOSPC2875L-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Difluorophenyl isothiocyanate 97%
Catalog # APOSPC2875L-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :171.17 g/mol
Boiling Pt:99 °C/30mmHg
Density:1.349 g/cm³ (25 °C)
Flash Pt:92 °C
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,III
 :GHS07
CAS Number:141106-52-7
Formula:C₇H₃F₂NS
MDL Number:MFCD00041044