Order Entry
Czech Republic
ContactUsLinkComponent
Bis[2-(fluorosulphonyl)tetrafluoroethyl]ether 98%
Corrosive
Bis[2-(fluorosulphonyl)tetrafluoroethyl]ether 98%
Catalog # APOSPC2855-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Bis[2-(fluorosulphonyl)tetrafluoroethyl]ether 98%
Catalog # APOSPC2855-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :382.15 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
 :GHS05
CAS Number:146829-79-0
Formula:C₄F₁₀O₅S₂
MDL Number:MFCD03094207