
2,4-Difluoro-N-methylaniline 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:138564-16-6
:
Formula:C₇H₇F₂N
MDL Number:MFCD00042472
:143.14 g/mol
Boiling Pt:94…96 °C/15mmHg
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together