

4-(Difluoromethoxy)toluene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1583-83-1
:
Formula:C₈H₈F₂O
MDL Number:MFCD00042253
:158.15 g/mol
Boiling Pt:50…52 °C/15mmHg
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together