Order Entry
Czech Republic
ContactUsLinkComponent
4-(Difluoromethoxy)iodobenzene 98%
Irritant
4-(Difluoromethoxy)iodobenzene 98%
Catalog # APOSPC2847R-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(Difluoromethoxy)iodobenzene 98%
Catalog # APOSPC2847R-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:217…219 °C
Density:1.871 g/cm³ (25 °C)
Flash Pt:110 °C
 :270.0153 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:128140-82-9
Formula:C₇H₅F₂IO
MDL Number:MFCD00085008