
4'-(Difluoromethoxy)acetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:83882-67-1
:
Formula:C₉H₈F₂O₂
MDL Number:MFCD00042250
:186.1600 g/mol
Boiling Pt:70…72 °C/0,5mmHg
Melting Pt:18…19 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together