1,3-Dimethyl-6-piperazino-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine 95%
Katalogové číslo APOSPC28231-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:299.3 g/mol
Storage Temperature:Ambient
CAS Number:952183-62-9
Formula:C₁₃H₁₆F₃N₅
MDL Number:MFCD09607951